CommentMaterial sourced in the U.S. and in other countries
Assay from Supplier's CofA≥97.5% (U.S. sourced material)
Refractive Index1.4500-1.4540 @ 20°C (non-U.S. sourced material)
Appearance (Color)Clear colorless
Assay (GC)≥97.5% (non-U.S. sourced material)
View more