(+/-)-Tetrahydrofurfuryl alcohol, 98%
(+/-)-Tetrahydrofurfuryl alcohol, 98%
(+/-)-Tetrahydrofurfuryl alcohol, 98%
Thermo Scientific Chemicals

(+/-)-Tetrahydrofurfuryl alcohol, 98%

CAS: 97-99-4 | C5H10O2 | 102.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2500 mL
Catalog number A17078.AP
also known as A17078-AP
Price (USD)
38.65
Online Exclusive
42.40
Save 3.75 (9%)
Each
Quantity:
500 mL
Request bulk or custom format
Price (USD)
38.65
Online Exclusive
42.40
Save 3.75 (9%)
Each
Chemical Identifiers
CAS97-99-4
IUPAC Name(oxolan-2-yl)methanol
Molecular FormulaC5H10O2
InChI KeyBSYVTEYKTMYBMK-UHFFFAOYNA-N
SMILESOCC1CCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
CommentMaterial sourced in the U.S. and in other countries
Assay from Supplier's CofA≥97.5% (U.S. sourced material)
Refractive Index1.4500-1.4540 @ 20°C (non-U.S. sourced material)
Appearance (Color)Clear colorless
Assay (GC)≥97.5% (non-U.S. sourced material)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only