3-Indoxyl sulfate potassium salt, 97%
3-Indoxyl sulfate potassium salt, 97%
3-Indoxyl sulfate potassium salt, 97%
Thermo Scientific Chemicals

3-Indoxyl sulfate potassium salt, 97%

CAS: 2642-37-7 | C8H6KNO4S | 251.297 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.25 g
1 g
5 g
Catalog number A17079.01
also known as A17079-01
Price (USD)
73.80
Each
Quantity:
0.25 g
Request bulk or custom format
Price (USD)
73.80
Each
Chemical Identifiers
CAS2642-37-7
IUPAC Namepotassium 1H-indol-3-yl sulfate
Molecular FormulaC8H6KNO4S
InChI KeyMDAWATNFDJIBBD-UHFFFAOYSA-M
SMILES[K+].[O-]S(=O)(=O)OC1=CNC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystalline powder
Assay (HPLC)≥96% (from supplier's CofA)
Assay from Supplier's CofA≥96.0%
Water Content (Karl Fischer Titration)≤2.0% (from supplier's CofA)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only