2-Chloronicotinic acid, 99%
2-Chloronicotinic acid, 99%
2-Chloronicotinic acid, 99%
Thermo Scientific Chemicals

2-Chloronicotinic acid, 99%

CAS: 2942-59-8 | C6H4ClNO2 | 157.553 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A17082.09
also known as A17082-09
Price (USD)
25.65
Special offer
Online exclusive
Ends: 30-Jun-2026
30.30
Save 4.65 (15%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
25.65
Special offer
Online exclusive
Ends: 30-Jun-2026
30.30
Save 4.65 (15%)
Each
Chemical Identifiers
CAS2942-59-8
IUPAC Name2-chloropyridine-3-carboxylic acid
Molecular FormulaC6H4ClNO2
InChI KeyIBRSSZOHCGUTHI-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CN=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Assay (HPLC)≥98.5%
Appearance (Color)White to cream
FormPowder
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only