N,N'-Diisopropylethylenediamine, 97%
N,N'-Diisopropylethylenediamine, 97%
N,N'-Diisopropylethylenediamine, 97%
Thermo Scientific™

N,N'-Diisopropylethylenediamine, 97%

CAS: 4013-94-9 | C8H20N2 | 144.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A17114.14
also known as A17114-14
Price (USD)
211.65
Online exclusive
235.00
Save 23.35
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
211.65
Online exclusive
235.00
Save 23.35
Each
Chemical Identifiers
CAS4013-94-9
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Refractive Index1.4255-1.4305 @ 20?C
Appearance (Color)Clear colorless
FormLiquid
N,N'-Diethylethylenediamine is used in the preparation of 2-imidazolidinecarboxaldehydes and 1,4,6,9-tetraalkyl-1,4,6,9-tetraaza-5,10-dioxaperhydroanthracenes by reacting with glyoxal. It acts as a ligand and form coordination complexes with copper(II), which shows thermochromic properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N′-Diethylethylenediamine is used in the preparation of 2-imidazolidinecarboxaldehydes and 1,4,6,9-tetraalkyl-1,4,6,9-tetraaza-5,10-dioxaperhydroanthracenes by reacting with glyoxal. It acts as a ligand and form coordination complexes with copper(II), which shows thermochromic properties.

Solubility
Miscible with water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only

General References:

  1. Yuan, Y.; Gong, F.; Cao, Y.; Chen, W.; Cheng, D.; Shuai, X. Biodegradable Multiamine Polymeric Vector for siRNA Delivery. J. Biomed. Nanotechnol. 2015, 11 (4), 668-679.
  2. Zhu, Y. J.; Chen, F. pH-Responsive Drug-Delivery Systems. Chem. Asian J. 2015, 10 (2), 284-305.