o-Dianisidine dihydrochloride, 98%
o-Dianisidine dihydrochloride, 98%
o-Dianisidine dihydrochloride, 98%
Thermo Scientific Chemicals

o-Dianisidine dihydrochloride, 98%

CAS: 20325-40-0 | C14H18Cl2N2O2 | 317.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A17175.06
also known as A17175-06
Price (USD)
159.65
Online Exclusive
177.00
Save 17.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
159.65
Online Exclusive
177.00
Save 17.35 (10%)
Each
Chemical Identifiers
CAS20325-40-0
IUPAC Namedihydrogen 3,3'-dimethoxy-[1,1'-biphenyl]-4,4'-diamine dichloride
Molecular FormulaC14H18Cl2N2O2
InChI KeyUXTIAFYTYOEQHV-UHFFFAOYSA-N
SMILES[H+].[H+].[Cl-].[Cl-].COC1=CC(=CC=C1N)C1=CC=C(N)C(OC)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to grey to purple-grey
FormCrystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5% (Dry wt. basis)
Loss on Drying≤5%
It is suitable for use in ELISA procedures. In neurohistochemistry o-Dianisidine dihydrochloride has demonstrated perikaryal labeling as well as retrograde and anterograde transport to the primary olfactory cortex. It has also been used for a paper based-micro fluidic device to semi-quantitatively measure lactate, uric acid, and glucose. 3,3'-Dimethoxybenzidine is used principally as an intermediate in the production of commercial bisazobiphenyl dyes for coloring textiles, paper, plastic, rubber, and leather.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is suitable for use in ELISA procedures. In neurohistochemistry o-Dianisidine dihydrochloride has demonstrated perikaryal labeling as well as retrograde and anterograde transport to the primary olfactory cortex. It has also been used for a paper based-micro fluidic device to semi-quantitatively measure lactate, uric acid, and glucose. 3,3′-Dimethoxybenzidine is used principally as an intermediate in the production of commercial bisazobiphenyl dyes for coloring textiles, paper, plastic, rubber, and leather.

Solubility
Soluble in water (50 mg/ml). It dissolves in ethyl alcohol ,acid and in organic solvent.

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Yanfen Peng.; Zhiqiang Gao.Amplified detection of microRNA based on ruthenium oxide nanoparticle-initiated deposition of an insulating film.Anal Chem.2011,83(3), 820-827.
  2. Wei Shen.; Huimin Deng.; Yuqian Ren, Zhiqiang Gao. A label-free microRNA biosensor based on DNAzyme-catalyzed and microRNA-guided formation of a thin insulating polymer film.Biosens Bioelectron.2013,44,171-176 .