Menthofuran, 95%
Menthofuran, 95%
Menthofuran, 95%
Thermo Scientific Chemicals

Menthofuran, 95%

CAS: 494-90-6 | C10H14O | 150.221 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A17192.03
also known as A17192-03
Price (USD)
50.00
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
50.00
Each
Chemical Identifiers
CAS494-90-6
IUPAC Name(6R)-3,6-dimethyl-4,5,6,7-tetrahydro-1-benzofuran
Molecular FormulaC10H14O
InChI KeyYGWKXXYGDYYFJU-SSDOTTSWSA-N
SMILESC[C@@H]1CCC2=C(C1)OC=C2C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Assay (GC)≥94.0%
Refractive Index1.4820-1.4880 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only