4-(Trifluoromethoxy)aniline, 98%
4-(Trifluoromethoxy)aniline, 98%
4-(Trifluoromethoxy)aniline, 98%
4-(Trifluoromethoxy)aniline, 98%
Thermo Scientific Chemicals

4-(Trifluoromethoxy)aniline, 98%

CAS: 461-82-5 | C7H6F3NO | 177.126 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A17208.14
also known as A17208-14
Price (USD)
107.65
Online Exclusive
119.00
Save 11.35 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
107.65
Online Exclusive
119.00
Save 11.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS461-82-5
IUPAC Name4-(trifluoromethoxy)aniline
Molecular FormulaC7H6F3NO
InChI KeyXUJFOSLZQITUOI-UHFFFAOYSA-N
SMILESNC1=CC=C(OC(F)(F)F)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Refractive Index1.4605-1.4655 @ 20?C
FormLiquid
Appearance (Color)Clear colorless to yellow or pale brown
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only