MOPS sodium salt, 97%
MOPS sodium salt, 97%
MOPS sodium salt, 97%
Thermo Scientific Chemicals

MOPS sodium salt, 97%

CAS: 71119-22-7 | C7H14NNaO4S | 231.242 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A17214.14
also known as A17214-14
Price (USD)
46.20
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
46.20
Each
Chemical Identifiers
CAS71119-22-7
IUPAC Namesodium 3-(morpholin-4-yl)propane-1-sulfonate
Molecular FormulaC7H14NNaO4S
InChI KeyMWEMXEWFLIDTSJ-UHFFFAOYSA-M
SMILES[Na+].[O-]S(=O)(=O)CCCN1CCOCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms (non-U.S. specification)
Acid-base back titration≥96.0 to ≤104.0% (non-U.S. specification)
Assay from Suppliers CofA≥96.0% (U.S. specification)
CommentSpecification differs for U.S. and non-U.S. material where indicated
Appearance (Color)White to pale cream
View more
3-(N-Morpholino)propanesulfonic Acid, Na; an analog to MES used in polyacrylamide gel electrophoresis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(N-Morpholino)propanesulfonic Acid, Na; an analog to MES used in polyacrylamide gel electrophoresis.

Solubility
Soluble in water (523 g/L at 20°C).

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
RUO – Research Use Only

General References:

  1. Munessar Sankar.; Roger G. Bates. Buffers for the physiological pH range: thermodynamic constants of 3-(N-morpholino)propanesulfonic acid from 5 to 50.degree.C.Anal. Chem.,1978,50(13), 1922-1924.
  2. K M Pan.; M Baldwin.; J Nguyen.; M Gasset.; A Serban.; D Groth.; I Mehlhorn.; Z Huang.; R J Fletterick.; and F E Cohen. Conversion of alpha-helices into beta-sheets features in the formation of the scrapie prion proteins.Proc. Natl. Acad. Sci.1993,90,(23), 10962-10966.