4-Nitrophenyl phenyl sulfide, 98%
4-Nitrophenyl phenyl sulfide, 98%
4-Nitrophenyl phenyl sulfide, 98%
Thermo Scientific Chemicals

4-Nitrophenyl phenyl sulfide, 98%

CAS: 952-97-6 | C12H9NO2S | 231.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A17278.06
also known as A17278-06
Price (USD)
63.65
Special offer
Online exclusive
Ends: 30-Jun-2026
75.10
Save 11.45 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
63.65
Special offer
Online exclusive
Ends: 30-Jun-2026
75.10
Save 11.45 (15%)
Each
Chemical Identifiers
CAS952-97-6
IUPAC Name1-nitro-4-(phenylsulfanyl)benzene
Molecular FormulaC12H9NO2S
InChI KeyRJCBYBQJVXVVKB-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(SC2=CC=CC=C2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)51.0-57.0?C
FormCrystals or powder or crystalline powder
Appearance (Color)Brown
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only