1-Decanol, 98+%
1-Decanol, 98+%
1-Decanol, 98+%
Thermo Scientific Chemicals

1-Decanol, 98+%

CAS: 112-30-1 | C10H22O | 158.285 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2500 mL
Catalog number A17288.0F
also known as A17288-0F
Price (USD)
106.00
Each
Quantity:
2500 mL
Request bulk or custom format
Price (USD)
106.00
Each
Chemical Identifiers
CAS112-30-1
IUPAC Namedecan-1-ol
Molecular FormulaC10H22O
InChI KeyMWKFXSUHUHTGQN-UHFFFAOYSA-N
SMILESCCCCCCCCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥97.5%
CommentSpecification differs for U.S. and non-U.S. material where indicated
Identification (FTIR)Conforms (non-U.S. specification)
View more
1-Decanol is used in the production of plasticizers, lubricants, surfactants and solvents. It is also used to study the thermal properties of polymer-monolithic stationary phases. Further, it is used to enhance homomeric glycine receptor function. In addition to this, it is used in daily flavor, food flavor and cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Decanol is used in the production of plasticizers, lubricants, surfactants and solvents. It is also used to study the thermal properties of polymer-monolithic stationary phases. Further, it is used to enhance homomeric glycine receptor function. In addition to this, it is used in daily flavor, food flavor and cosmetics.

Solubility
Soluble in alcohol, ether. Insoluble in water.Miscible with carbon tetrachloride, ethanol, ether, acetone, benzene, chloroform and glacial acetic acid. Immiscible with water.

Notes
Incompatible with strong oxidizing agents, acid chlorides and acid anhydrides.
RUO – Research Use Only

General References:

  1. Zhang, M.; Zhao, Y.; Liu, Q.; Yang, L.; Fan, G.; Li, F. A La-doped Mg-Al mixed metal oxide supported copper catalyst with enhanced catalytic performance in transfer dehydrogenation of 1-decanol. Dalton Trans. 2016, 45 (3), 1093-1102.
  2. Echaroj, S.; Santikunaporn, M.; Chavadej, S. Micro-kinetic modeling of the catalytic dehydration of 1-decanol over precipitated gamma-Al2O3. Reac. Kinet. Mech. Cat. 2015, 114 (1), 75-91.