1,5-Diiodopentane, 97%, stab. with copper
1,5-Diiodopentane, 97%, stab. with copper
1,5-Diiodopentane, 97%, stab. with copper
Thermo Scientific Chemicals

1,5-Diiodopentane, 97%, stab. with copper

CAS: 628-77-3 | C5H10I2 | 323.944 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A17298.14
also known as A17298-14
Price (USD)
53.65
Online Exclusive
59.20
Save 5.55 (9%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
53.65
Online Exclusive
59.20
Save 5.55 (9%)
Each
Chemical Identifiers
CAS628-77-3
IUPAC Name1,5-diiodopentane
Molecular FormulaC5H10I2
InChI KeyIAEOYUUPFYJXHN-UHFFFAOYSA-N
SMILESICCCCCI
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colourless or yellow to orange
Refractive Index1.5980 - 1.6040 @20?C
Assay (GC)> 96.0%
Formliquid
1,5-Diiodopentane is used as a crosslinking reagent. It is used in the preparation of (iodoalkylaminocarbene)tungsten complexes. It is also used as alkylation to form unsubstituted cyclohexane. It is involved in the preparation of poly(1-vinyl imidazole-co-elthylene glycol methyl ether acrylate electrolytes and used for quasi-solid state dye-sensitized solar cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,5-Diiodopentane is used as a crosslinking reagent. It is used in the preparation of (iodoalkylaminocarbene)tungsten complexes. It is also used as alkylation to form unsubstituted cyclohexane. It is involved in the preparation of poly(1-vinyl imidazole-co-elthylene glycol methyl ether acrylate electrolytes and used for quasi-solid state dye-sensitized solar cells.

Solubility
Immiscible with water.

Notes
Light sensitive. Incompatible with strong oxidizing agents, strong acids and strong bases.
RUO – Research Use Only

General References:

  1. Amata, E.; Bland, N. D.; Campbell, R. K.; Pollastri, M. P. Evaluation of pyrrolidine and pyrazolone derivatives as inhibitors of trypanosomal phosphodiesterase B1 (TbrPDEB1). Tetrahedron Lett. 2015, 56 (21), 2832-2835.
  2. Deng, R.; Liang, F.; Qu, X.; Wang, Q.; Zhu, J.; Yang, Z. Diblock Copolymer Based Janus Nanoparticles. Macromolecules 2015, 48 (3), 750-755.