Isopropyl trifluoroacetate, 97%
Isopropyl trifluoroacetate, 97%
Isopropyl trifluoroacetate, 97%
Isopropyl trifluoroacetate, 97%
Thermo Scientific Chemicals

Isopropyl trifluoroacetate, 97%

CAS: 400-38-4 | C5H7F3O2 | 156.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A17322.22
also known as A17322-22
Price (USD)
132.65
Online Exclusive
147.00
Save 14.35 (10%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
132.65
Online Exclusive
147.00
Save 14.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS400-38-4
IUPAC Namepropan-2-yl 2,2,2-trifluoroacetate
Molecular FormulaC5H7F3O2
InChI KeyASAXRKSDVDALDT-UHFFFAOYSA-N
SMILESCC(C)OC(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow or pink
FormLiquid
Assay (GC)≥96.0%
Identification (FTIR)Conforms
Refractive Index1.3165-1.3215 @ 20°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only