3-Acetyl-2,5-dimethylthiophene, 99%
Thermo Scientific Chemicals

3-Acetyl-2,5-dimethylthiophene, 99%

CAS: 2530-10-1 | C9H22O3Si | 206.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number A17329.09
also known as A17329-09
Price (USD)
44.65
Online Exclusive
49.10
Save 4.45 (9%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
44.65
Online Exclusive
49.10
Save 4.45 (9%)
Each
Chemical Identifiers
CAS2530-10-1
IUPAC Name1-(2,5-dimethylthiophen-3-yl)ethan-1-one
Molecular FormulaC8H10OS
InChI KeyPUSJAEJRDNPYKM-UHFFFAOYSA-N
SMILESCC(=O)C1=C(C)SC(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear pale yellow to yellow to brown
Refractive Index1.5435-1.5465 @ 20?C
Assay (GC)≥98.5%
It is used in the synthesis of heterocyclic ketimines, 3-acetyl-2,5-dimethylthiophene thiosemicarbazone and 3-acetyl-2,5- dimethylthiophene semicarbazone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis of heterocyclic ketimines, 3-acetyl-2,5-dimethylthiophene thiosemicarbazone and 3-acetyl-2,5- dimethylthiophene semicarbazone.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Krishna Sharma, et. al. Microwave assisted synthesis, characterization and biological evaluation of palladium and platinum complexes with azomethines.Spectrochim Acta A Mol Biomol Spectrosc.,2010,75(1), 422-427.
  2. Janine M. Cooney, et al. Ortho-manganated arenes in synthesis: IV.Ortho-manganation of substituted acetophenones and of heteroaromatic methyl ketones. The crystal structures of two cyclometallated acetylthiophene derivatives.J. Organomet. Chem.,1988,349(1-2), 197-207.