alpha-Bromo-gamma-butyrolactone, 97%
alpha-Bromo-gamma-butyrolactone, 97%
alpha-Bromo-gamma-butyrolactone, 97%
Thermo Scientific Chemicals

alpha-Bromo-gamma-butyrolactone, 97%

CAS: 5061-21-2 | C4H5BrO2 | 164.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A17344.22
also known as A17344-22
Price (USD)
117.65
Online Exclusive
130.00
Save 12.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
117.65
Online Exclusive
130.00
Save 12.35 (10%)
Each
Chemical Identifiers
CAS5061-21-2
IUPAC Name3-bromooxolan-2-one
Molecular FormulaC4H5BrO2
InChI KeyLFJJGHGXHXXDFT-UHFFFAOYNA-N
SMILESBrC1CCOC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow or pale brown
FormLiquid
Assay (GC)≥95.0%
Identification (FTIR)Conforms
Refractive Index1.5055-1.5105 @ 20°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only