2-Ethylpyridine, 98%
2-Ethylpyridine, 98%
2-Ethylpyridine, 98%
Thermo Scientific Chemicals

2-Ethylpyridine, 98%

CAS: 100-71-0 | C7H9N | 107.156 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number A17352.22
also known as A17352-22
Price (USD)
163.65
Online Exclusive
182.00
Save 18.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
163.65
Online Exclusive
182.00
Save 18.35 (10%)
Each
Chemical Identifiers
CAS100-71-0
IUPAC Name2-ethylpyridine
Molecular FormulaC7H9N
InChI KeyNRGGMCIBEHEAIL-UHFFFAOYSA-N
SMILESCCC1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4955-1.4995 @ 20?C
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥97.5%
FormLiquid
2-Ethylpyridine is used to study its effect on the proliferation and survival of human umbilical vein endothelial cells (HUVECs), HMVECs from lung, and NIH 3T3 cells. 2-Ethylpyridine linked with silica is a popular stationary phase for chiral and achiral separations in supercritical fluid chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethylpyridine is used to study its effect on the proliferation and survival of human umbilical vein endothelial cells (HUVECs), HMVECs from lung, and NIH 3T3 cells. 2-Ethylpyridine linked with silica is a popular stationary phase for chiral and achiral separations in supercritical fluid chromatography.

Solubility
Miscible with water (42 g/L at20°C).

Notes
Store at room temperature. Keep away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. María L. Buil.; Miguel A. Esteruelas.; Karin Garcés.; Montserrat Oliván.; Enrique Oñate. Understanding the Formation of N-H Tautomers from α-Substituted Pyridines: Tautomerization of 2-Ethylpyridine Promoted by Osmium. J. Am. Chem. Soc. 2007, 129 (36),10998-10999.
  2. M. Goldstein, E.F. Mooney.; A. Anderson.; H.A. Gebbie. The infrared spectra and structure of complexes of copper(II) halides and heterocyclic bases. Spectrochim. Acta. 1965, 21 (1),105-117.