N,N-Dimethylhexylamine, 99%
N,N-Dimethylhexylamine, 99%
N,N-Dimethylhexylamine, 99%
N,N-Dimethylhexylamine, 99%
Thermo Scientific Chemicals

N,N-Dimethylhexylamine, 99%

CAS: 4385-04-0 | C8H19N | 129.247 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A17364.14
also known as A17364-14
Price (USD)
182.00
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
182.00
Each
Add to cart
Chemical Identifiers
CAS4385-04-0
IUPAC Namehexyldimethylazanium
Molecular FormulaC8H20N
InChI KeyQMHNQZGXPNCMCO-UHFFFAOYSA-O
SMILESCCCCCC[NH+](C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Refractive Index1.4120-1.4150 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥98.5%
FormLiquid
N,N-Dimethylhexylamine is used as an ion-pairing agent to study the poor retention of uridine diphosphate-linked intermediates on reverse phase media. It is also used in the determination of five monophosphate nucleotides in baby foods. It acts as an additive and encapsulated into the nanoparticle with salinomycin (sali), which has selective toxicity to cancer stem cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethylhexylamine is used as an ion-pairing agent to study the poor retention of uridine diphosphate-linked intermediates on reverse phase media. It is also used in the determination of five monophosphate nucleotides in baby foods. It acts as an additive and encapsulated into the nanoparticle with salinomycin (sali), which has selective toxicity to cancer stem cells.

Solubility
Miscible with methanol.

Notes
Incompatible with strong acids and strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Guyon, C.; Da Silva, E.; Lafon, R.; Métay, E.; Lemaire, M. Reductive amination using a combination of CaH2 and noble metal. RSC Adv. 2015, 5 (3), 2292-2298.
  2. Arges, C. G.; Wang, L.; Jung, M. S.; Ramani, V. Mechanically Stable Poly(arylene ether) Anion Exchange Membranes Prepared from Commercially Available Polymers for Alkaline Electrochemical Devices. J. Electrochem. Soc. 2015, 162 (7), F686-F693.