Ethyl 4-acetyl-3,5-dimethylpyrrole-2-carboxylate, 98%
Ethyl 4-acetyl-3,5-dimethylpyrrole-2-carboxylate, 98%
Ethyl 4-acetyl-3,5-dimethylpyrrole-2-carboxylate, 98%
Thermo Scientific Chemicals

Ethyl 4-acetyl-3,5-dimethylpyrrole-2-carboxylate, 98%

CAS: 2386-26-7 | C11H15NO3 | 209.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A17365.09
also known as A17365-09
Price (USD)
79.65
Online exclusive
88.00
Save 8.35
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
79.65
Online exclusive
88.00
Save 8.35
Each
Chemical Identifiers
CAS2386-26-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale orange
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Melting Point (clear melt)139.0-145.0°C
It is utilized as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is utilized as pharmaceutical intermediate.

Solubility
Slightly soluble in water. 

Notes
Refrigerated (2-4°C). Keep away from strong oxidizing agents and bases.
RUO – Research Use Only

General References:

  1. Michael T. Huggins.; Adrianne K. Tipton.; Qingqi Chen.; David A. Lightner. Hydrogen-Bonded Double Strands: Crystal Structure and Spectroscopic Propertiesof a 2,2'-Dipyrryl Ketone.Monatsh. Chem.. 2000, 131 (8), 825-838.
  2. Ulli Eisner.; P. H. Gore. The light absorption of pyrroles. Part I. Ultraviolet spectra.J. Chem. Soc. 1958, (0), 922-927.