N-Benzyl-N-phenylhydrazine hydrochloride, 98+%
N-Benzyl-N-phenylhydrazine hydrochloride, 98+%
N-Benzyl-N-phenylhydrazine hydrochloride, 98+%
N-Benzyl-N-phenylhydrazine hydrochloride, 98+%
Thermo Scientific Chemicals

N-Benzyl-N-phenylhydrazine hydrochloride, 98+%

CAS: 5705-15-7 | C13H15ClN2 | 234.73 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A17367.06
also known as A17367-06
Price (USD)
46.65
Online Exclusive
51.80
Save 5.15 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
46.65
Online Exclusive
51.80
Save 5.15 (10%)
Each
Add to cart
Chemical Identifiers
CAS5705-15-7
IUPAC Namehydrogen 1-benzyl-1-phenylhydrazine chloride
Molecular FormulaC13H15ClN2
InChI KeyJTYLHYOCBGPMNO-UHFFFAOYSA-N
SMILES[H+].[Cl-].NN(CC1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White to pale brown
Assay (Titration ex Chloride)≥98.0 to ≤102.0%
Melting Point (clear melt)167-181?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only