2,6-Difluorobenzyl bromide, 96%
2,6-Difluorobenzyl bromide, 96%
2,6-Difluorobenzyl bromide, 96%
Thermo Scientific Chemicals

2,6-Difluorobenzyl bromide, 96%

CAS: 85118-00-9 | C7H5BrF2 | 207.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A17434.06
also known as A17434-06
Price (USD)
55.65
Online Exclusive
61.60
Save 5.95 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
55.65
Online Exclusive
61.60
Save 5.95 (10%)
Each
Chemical Identifiers
CAS85118-00-9
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥95.0%
Appearance (Color)White to pale cream to pale yellow
Melting Point (clear melt)47.0-57.0°C
FormCrystals or powder or crystalline powder or lumps or fused solid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only