Methyl Red sodium salt
Methyl Red sodium salt
Methyl Red sodium salt
Thermo Scientific Chemicals

Methyl Red sodium salt

CAS: 845-10-3 | C15H14N3NaO2 | 291.286 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A17455.36
also known as A17455-36
Price (USD)
235.65
Special offer
Online exclusive
Ends: 28-Dec-2026
277.00
Save 41.35
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
235.65
Special offer
Online exclusive
Ends: 28-Dec-2026
277.00
Save 41.35
Each
Chemical Identifiers
CAS845-10-3
IUPAC Namesodium 2-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzoate
Molecular FormulaC15H14N3NaO2
InChI KeyGNTPCYMJCJNRQB-UHFFFAOYSA-M
SMILES[Na+].CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Specific Absorbance (E 1%/1cm)≥585 (@ lambda max)
Appearance (Color)Orange to Brown
Appearance (Color)pH4.2 (Violet Red) - pH6.2 (Brown Yellow)
Solution Test0.1% w/v (in distilled water): Clear
lambda-max (UV-Vis)430.0-440.0nm (in distilled water)
View more
Methyl red is an indicator dye that turns red in acidic solutions. It is a pH indicator, turning red under pH 4.4, yellow over pH 6.2, and orange in between those values. It is used in microbiology in the Voges-Proskauer test to identify bacteria that produce stable acids through mixed acid fermentation of glucose. Methyl-red positive bacteria include Proteus vulgaris and Escherichia coli. Methyl-red negative bacteria include Enterobacter aerogenes and Serratia marcescens.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl red is an indicator dye that turns red in acidic solutions. It is a pH indicator, turning red under pH 4.4, yellow over pH 6.2, and orange in between those values. It is used in microbiology in the Voges-Proskauer test to identify bacteria that produce stable acids through mixed acid fermentation of glucose. Methyl-red positive bacteria include Proteus vulgaris and Escherichia coli. Methyl-red negative bacteria include Enterobacter aerogenes and Serratia marcescens.

Solubility
Soluble in water (800 mg/ml at 25°C), and slightly soluble in ethanol at (25°C).

Notes
Store at room temperature. Reactive with oxidizing agents.
RUO – Research Use Only

General References:

  1. Bin Zhao.; Jeffrey S. Moore.; David J. Beebe. Surface-Directed Liquid Flow Inside Microchannels. Science. 2001, 291 (5506),1023-1026.
  2. Shizhuo Yin.; Kun-Wook Chung.; Xin Zhu. A novel all-optic tunable long-period grating using a unique double-cladding layer. Opt. Commun.. 2001, 196 (1-6),181-186.