2-Nitrothiophene, tech. 85%
2-Nitrothiophene, tech. 85%
2-Nitrothiophene, tech. 85%
Thermo Scientific Chemicals

2-Nitrothiophene, tech. 85%

CAS: 609-40-5 | C4H3NO2S | 129.133 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A17464.06
also known as A17464-06
Price (USD)
41.65
Online Exclusive
46.00
Save 4.35 (9%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
41.65
Online Exclusive
46.00
Save 4.35 (9%)
Each
Chemical Identifiers
CAS609-40-5
IUPAC Name2-nitrothiophene
Molecular FormulaC4H3NO2S
InChI KeyJIZRGGUCOQKGQD-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder and/or lumps/lumpy powder
CommentPale material darkens with time, colour may become non-uniform
Appearance (Color)Pale cream to brown or yellow to orange or cream/orange
Assay (GC)≥83.0%
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only