2-Diethylaminoethyl 4-aminobenzoate hydrochloride, 99%
2-Diethylaminoethyl 4-aminobenzoate hydrochloride, 99%
2-Diethylaminoethyl 4-aminobenzoate hydrochloride, 99%
Thermo Scientific Chemicals

2-Diethylaminoethyl 4-aminobenzoate hydrochloride, 99%

CAS: 51-05-8 | C13H21ClN2O2 | 272.77 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A17485.30
also known as A17485-30
Price (USD)
100.65
Online Exclusive
112.00
Save 11.35 (10%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
100.65
Online Exclusive
112.00
Save 11.35 (10%)
Each
Chemical Identifiers
CAS51-05-8
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)153.0-159.0°C
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5% (non-U.S. specification)
Assay (Titration ex Chloride)≥98.5 to ≤101.5% (non-U.S. specification)
View more
A sodium channel blocker

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only