Search
Search
Suitable for battery materials development.
Applications
Sodium chromate tetrahydrate is used as a corrosion inhibitor, wood preservative and an auxiliary in textile industry. It serves as an oxidant, which is used to prepare carboxylic acids and ketones from primary and secondary alcohols respectively. Further, it is used as a starting material to prepare other chromium compounds and as a catalyst in organic synthesis especially pharmaceuticals and agro chemicals.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible withstrong reducing agents, organic materials, strong acids, combustible materials and powdered metals.
General References: