Azure I
Azure I
Azure I
Thermo Scientific Chemicals

Azure I

CAS: 531-55-5 | C15H16ClN3S | 305.824 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A17508.06
also known as A17508-06
Price (USD)
44.90
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
44.90
Each
Chemical Identifiers
CAS531-55-5
IUPAC Name3-(dimethylamino)-7-(methylamino)-5λ⁴-phenothiazin-5-ylium chloride
Molecular FormulaC15H16ClN3S
InChI KeyKFZNPGQYVZZSNV-UHFFFAOYSA-M
SMILES[Cl-].CNC1=CC=C2N=C3C=CC(=CC3=[S+]C2=C1)N(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Specific Absorbance (E 1%/1cm)>1250 (at lambda-max)
FormPowder
lambda-max (UV-Vis)645 - 650nm (aqueous solution)
Appearance (Color)Dark green or brown to black
Loss on Drying<8% (105?C, 1 hour)
View more
Azure B is used for staining semi-thin sections of plant tissue. This dye is also useful in the NCCLS method for blood smears and Lillie's modified Nocht's method for paraffin sections.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Azure B is used for staining semi-thin sections of plant tissue. This dye is also useful in the NCCLS method for blood smears and Lillie′s modified Nocht′s method for paraffin sections.

Solubility
Soluble in 1 mg/ml water. Sparingly soluble in alcohol.

Notes
Store in a cool place. Ensure proper ventilation. Keep away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Chen-Xin Cai.; Kuan-Hong Xue. Electrocatalysis of NADH oxidation with electropolymerized films of azure I. J. Electroanal. Chem. 1997, 427 (1-2), 147-153.
  2. Na Li .; Ruo Yuan.; Yaqin Chai.; Shihong Chen.; Haizhen An.; Wenjuan Li. New Antibody Immobilization Strategy Based on Gold Nanoparticles and Azure I/Multi-Walled Carbon Nanotube Composite Membranes for an Amperometric Enzyme Immunosensor. J. Phys. Chem. 2007, 111 (24),8443-8450.