(+)-Cinchonine, 98+%, cont. up to 3% quinidine/dihydroquinidine and 3% quinine/dihydroquinine
(+)-Cinchonine, 98+%, cont. up to 3% quinidine/dihydroquinidine and 3% quinine/dihydroquinine
(+)-Cinchonine, 98+%, cont. up to 3% quinidine/dihydroquinidine and 3% quinine/dihydroquinine
Thermo Scientific Chemicals

(+)-Cinchonine, 98+%, cont. up to 3% quinidine/dihydroquinidine and 3% quinine/dihydroquinine

CAS: 118-10-5 | C19H22N2O | 294.40 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A17523.22
also known as A17523-22
Price (USD)
122.65
Online Exclusive
136.00
Save 13.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
122.65
Online Exclusive
136.00
Save 13.35 (10%)
Each
Chemical Identifiers
CAS118-10-5
IUPAC Name(S)-[(2R,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](quinolin-4-yl)methanol
Molecular FormulaC19H22N2O
InChI KeyKMPWYEUPVWOPIM-FRYPIZGFNA-N
SMILES[H][C@@]1(CN2CCC1C[C@]2([H])[C@@H](O)C1=CC=NC2=CC=CC=C12)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Optical Rotation260? ? 9? (c=2, 0.2M HCl)
Assay (Non-aqueous acid-base Titration)≥98.0 to ≤102.0%
Impurity contentQuinine/Dihydroquinine ≤3.0%
Impurity contentQuinidine/Dihydroquinidine ≤3.0%
Appearance (Color)White to pale cream
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only