Acetoacetanilide, 98+%
Acetoacetanilide, 98+%
Acetoacetanilide, 98+%
Acetoacetanilide, 98+%
Thermo Scientific Chemicals

Acetoacetanilide, 98+%

CAS: 102-01-2 | C10H11NO2 | 177.203 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number A17534.0B
also known as A17534-0B
Price (USD)
55.70
Each
Add to cart
Quantity:
1000 g
Request bulk or custom format
Price (USD)
55.70
Each
Add to cart
Chemical Identifiers
CAS102-01-2
IUPAC Name3-oxo-N-phenylbutanamide
Molecular FormulaC10H11NO2
InChI KeyDYRDKSSFIWVSNM-UHFFFAOYSA-N
SMILESCC(=O)CC(=O)NC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream or pale grey
Assay (HPLC)≥98.0%
Melting Point81.5-87.5?C
Identification (FTIR)Conforms
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only