Polyethylene glycol 6,000
Polyethylene glycol 6,000
Polyethylene glycol 6,000
Thermo Scientific Chemicals

Polyethylene glycol 6,000

CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
5000 g
Catalog number A17541.0B
Price (USD)
76.65
Online exclusive
85.50
Save 8.85
Each
Quantity:
1000 g
Request bulk or custom format
Price (USD)
76.65
Online exclusive
85.50
Save 8.85
Each
Chemical Identifiers
CAS25322-68-3
Molecular Formula(C2H4O)n
InChI KeyLYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILES[H]OCCO
SpecificationsSpecification SheetSpecification Sheet
Viscosity200-270 mPas (50%, Water @ 20?C)
Appearance (Color)White to cream
CommentAverage mol weight 5400 - 6600 (based on OH value)
Melting Point (clear melt)55-63?C
pH4.5-7.5 (10%, Water)
View more
Polyethylene glycol is used in the modification of therapeutic proteins and peptides to enhance its solubility. It is also used in the fabrication of bioactive and immuno isolating barriers for encapsulation of cells. In Gas chromatorgraphy, it is used as a polar stationary phase. It is used as mold release agent and lubricant in fabricating elastomers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only