2-Fluoro-5-(trifluoromethyl)aniline, 97%
2-Fluoro-5-(trifluoromethyl)aniline, 97%
2-Fluoro-5-(trifluoromethyl)aniline, 97%
2-Fluoro-5-(trifluoromethyl)aniline, 97%
Thermo Scientific Chemicals

2-Fluoro-5-(trifluoromethyl)aniline, 97%

CAS: 535-52-4 | C7H5F4N | 179.118 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A17556.14
also known as A17556-14
Price (USD)
199.65
Online Exclusive
222.00
Save 22.35 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
199.65
Online Exclusive
222.00
Save 22.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS535-52-4
IUPAC Name2-fluoro-5-(trifluoromethyl)aniline
Molecular FormulaC7H5F4N
InChI KeyDRKWGMXFFCPZLW-UHFFFAOYSA-N
SMILESNC1=CC(=CC=C1F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow
FormLiquid
Refractive Index1.4585-1.4635 @ 20?C
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only