Cyclohexanol, 99%
Cyclohexanol, 99%
Cyclohexanol, 99%
Thermo Scientific Chemicals

Cyclohexanol, 99%

CAS: 108-93-0 | C6H12O | 100.161 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2500 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A17576.AE
also known as A17576-AE
Price (USD)
32.20
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
32.20
Each
Chemical Identifiers
CAS108-93-0
IUPAC Namecyclohexanol
Molecular FormulaC6H12O
InChI KeyHPXRVTGHNJAIIH-UHFFFAOYSA-N
SMILESOC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥98.5%
Identification (FTIR)Conforms
Cyclohexanol is used in making soaps, dry cleaning agents and plasticizers. It acts as a solvent for rubber, resins, nitrocellulose, metallic soaps, oils, esters and ethers. It is used in the preparation of adipic acid, hexamethylene diamine, cyclohexanone and caprolactam. It acts as a soap stabilizing agent, disinfection medicated soap and detergent emulsion. It is also useful for paint blending agent, leather degreasing agent and polishes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexanol is used in making soaps, dry cleaning agents and plasticizers. It acts as a solvent for rubber, resins, nitrocellulose, metallic soaps, oils, esters and ethers. It is used in the preparation of adipic acid, hexamethylene diamine, cyclohexanone and caprolactam. It acts as a soap stabilizing agent, disinfection medicated soap and detergent emulsion. It is also useful for paint blending agent, leather degreasing agent and polishes.

Solubility
Miscible with ethanol, ethyl ether acetone, chloroform, ethyl acetate, linseed oil and benzene. Slightly miscible with cold water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents, hydrogen peroxide and nitric acid.
RUO – Research Use Only

General References:

  1. Cyclohexyl esters can be used to protect carboxylic acids, including aspartate residues in peptide synthesis, resulting in reduced aspartimide formation during acidic or basic reactions: Synthesis, 361 (1992). They are relatively stable to TFA but can be readily cleaved by TfOH or HF: Int. J. Pept. Protein Res., 13, 418 (1979); Tetrahedron Lett., 4033 (1979); Chem. Pharm. Bull., 34, 864 (1986). See also 2,4-Dimethyl-3-pentanol, B20821 .
  2. Schemeth, D.; Noël, J. C.; Jakschitz, T.; Rainer, M.; Tessadri, R.; Huck, C. W.; Bonn, G. K. Poly(N-vinylimidazole/ethylene glycol dimethacrylate) for the purification and isolation of phenolic acids. Anal. Chim. Acta 2015, 885, 199-206.
  3. Zhang, P.; Deng, J.; Mao, J.; Li, H.; Wang, Y. Selective aerobic oxidation of alcohols by a mesoporous graphitic carbon nitride-hydroxyphthalimide system under visible-light illumination at room temperature. Chin. J. Catal. 2015, 36 (9), 1580-1586.