Nonenylsuccinic anhydride, 96%
Nonenylsuccinic anhydride, 96%
Nonenylsuccinic anhydride, 96%
Thermo Scientific Chemicals

Nonenylsuccinic anhydride, 96%

CAS: 28928-97-4 | C13H20O3
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number A17607.18
also known as A17607-18
Price (USD)
41.65
Online exclusive
46.20
Save 4.55
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
41.65
Online exclusive
46.20
Save 4.55
Each
Chemical Identifiers
CAS28928-97-4
IUPAC Name3-[(1E)-non-1-en-1-yl]oxolane-2,5-dione
Molecular FormulaC13H20O3
InChI KeyFGQUIQAGZLBOGL-CMDGGOBGNA-N
SMILESCCCCCCCC=CC1CC(=O)OC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥95.0 to ≤105.0%
Refractive Index1.4715-1.4775 @20?C
Appearance (Color)Clear colorless to pale brown
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only