Adenine hydrochloride, 98+%, cont. up to ca 5% water
Adenine hydrochloride, 98+%, cont. up to ca 5% water
Adenine hydrochloride, 98+%, cont. up to ca 5% water
Thermo Scientific Chemicals

Adenine hydrochloride, 98+%, cont. up to ca 5% water

CAS: 2922-28-3 | C5H6ClN5 | 171.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A17622.06
also known as A17622-06
Price (USD)
26.65
Online Exclusive
29.20
Save 2.55 (9%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
26.65
Online Exclusive
29.20
Save 2.55 (9%)
Each
Chemical Identifiers
CAS2922-28-3
IUPAC Name7H-purin-6-amine hydrochloride
Molecular FormulaC5H6ClN5
InChI KeyUQVDQSWZQXDUJB-UHFFFAOYSA-N
SMILESCl.NC1=C2NC=NC2=NC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration ex Chloride)>98.0 to <102.0% (Dry wt. basis)
Appearance (Color)White
Water Content (Karl Fischer Titration)< 7.5% (hemihydrate = 5.0%)
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only