5-Bromo-2-nitrobenzotrifluoride, 98+%
5-Bromo-2-nitrobenzotrifluoride, 98+%
5-Bromo-2-nitrobenzotrifluoride, 98+%
Thermo Scientific Chemicals

5-Bromo-2-nitrobenzotrifluoride, 98+%

CAS: 344-38-7 | C7H3BrF3NO2 | 270.005 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A17634.06
also known as A17634-06
Price (USD)
19.60
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
19.60
Each
Chemical Identifiers
CAS344-38-7
IUPAC Name4-bromo-1-nitro-2-(trifluoromethyl)benzene
Molecular FormulaC7H3BrF3NO2
InChI KeyZHLYHEDQTJZYFI-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(Br)C=C1C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
Appearance (Color)Yellow
FormCrystals or powder or crystalline powder or fused solid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only