Menthone, mixture of isomers, 98%
Menthone, mixture of isomers, 98%
Menthone, mixture of isomers, 98%
Thermo Scientific Chemicals

Menthone, mixture of isomers, 98%

CAS: 10458-14-7 | C10H18O | 154.253 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1000 g
Catalog number A17665.0B
also known as A17665-0B
Price (USD)
371.65
Special offer
Online exclusive
Ends: 28-Dec-2026
437.00
Save 65.35
Each
Quantity:
1000 g
Request bulk or custom format
Price (USD)
371.65
Special offer
Online exclusive
Ends: 28-Dec-2026
437.00
Save 65.35
Each
Chemical Identifiers
CAS10458-14-7
IUPAC Name5-methyl-2-(propan-2-yl)cyclohexan-1-one
Molecular FormulaC10H18O
InChI KeyNFLGAXVYCFJBMK-UHFFFAOYNA-N
SMILESCC(C)C1CCC(C)CC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥97.5% (sum of 2 components)
CompositionA mixture of L-menthone (predominantly) and D-isomenthone.
Identification (FTIR)Conforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only