3-Nitrobenzenesulfonyl chloride, 98%
3-Nitrobenzenesulfonyl chloride, 98%
3-Nitrobenzenesulfonyl chloride, 98%
3-Nitrobenzenesulfonyl chloride, 98%
Thermo Scientific Chemicals

3-Nitrobenzenesulfonyl chloride, 98%

CAS: 121-51-7 | C6H4ClNO4S | 221.611 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A17670.14
also known as A17670-14
Price (USD)
72.65
Online Exclusive
81.00
Save 8.35 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
72.65
Online Exclusive
81.00
Save 8.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS121-51-7
IUPAC Name3-nitrobenzene-1-sulfonyl chloride
Molecular FormulaC6H4ClNO4S
InChI KeyMWWNNNAOGWPTQY-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=CC(=C1)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to cream
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
Melting Point (clear melt)58.0-65.0?C
It is employed as a biochemical for proteomics research. It is also used in the preparation of: acyl-2-aminobenzimidazole analogs and 6-chloro-2-methyl-3-{1-[(3-nitrophenyl)sulfonyl]-1H-pyrazol-3-yl}imidazo[1,2-a]pyridine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a biochemical for proteomics research. It is also used in the preparation of: acyl-2-aminobenzimidazole analogs and 6-chloro-2-methyl-3-{1-[(3-nitrophenyl)sulfonyl]-1H-pyrazol-3-yl}imidazo[1,2-a]pyridine.

Solubility
Hydrolyzes slowly in water.

Notes
Moisture Sensitive. Store away from oxidizing agents, active metals and bases. Keep the container tightly closed in a cool, dry place.
RUO – Research Use Only

General References:

  1. Masahiko Hayakawa.; Hiroyuki Kaizawa.; Ken-Ichi Kawaguchi.; Noriko Ishikawa.; Tomonobu Koizumi.; Takahide Ohishi.; Mayumi Yamano.; Minoru Okada.; Mitsuaki Ohta.; Shin-Ichi Tsukamoto.; Florence I Raynaud.; Michael D Waterfield.; Peter Parker.; Paul Workman. Synthesis and biological evaluation of imidazo[1,2-a]pyridine derivatives as novel PI3 kinase p110alpha inhibitors.Bioorg Med Chem.2007,15(1), 403-412.
  2. Jay P Powers.; Shyun Li.; Juan C Jaen.; Jinqian Liu.; Nigel P C Walker.; Zhulun Wang.; Holger Wesche. Discovery and initial SAR of inhibitors of interleukin-1 receptor-associated kinase-4.Bioorg Med Chem Lett.2006,16(11), 2842-2845.