N-Benzyloxycarbonyl-L-glutamic acid, 99%
N-Benzyloxycarbonyl-L-glutamic acid, 99%
N-Benzyloxycarbonyl-L-glutamic acid, 99%
N-Benzyloxycarbonyl-L-glutamic acid, 99%
Thermo Scientific Chemicals

N-Benzyloxycarbonyl-L-glutamic acid, 99%

CAS: 1155-62-0 | C13H15NO6 | 281.264 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number A17686.18
also known as A17686-18
Price (USD)
104.65
Online Exclusive
116.00
Save 11.35 (10%)
Each
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)
104.65
Online Exclusive
116.00
Save 11.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS1155-62-0
IUPAC Name2-{[(benzyloxy)carbonyl]amino}pentanedioic acid
Molecular FormulaC13H15NO6
InChI KeyPVFCXMDXBIEMQG-UHFFFAOYNA-N
SMILESOC(=O)CCC(NC(=O)OCC1=CC=CC=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Melting Point (clear melt)114.0-120.0?C
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Optical Rotation-7.9 ± 0.1? (c=4 in acetic acid)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only