Heptanoic acid, 98+%
Heptanoic acid, 98+%
Heptanoic acid, 98+%
Thermo Scientific Chemicals

Heptanoic acid, 98+%

CAS: 111-14-8 | C7H14O2 | 130.187 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2500 mL
Catalog number A17704.AP
also known as A17704-AP
Price (USD)
65.65
Online Exclusive
73.20
Save 7.55 (10%)
Each
Quantity:
500 mL
Request bulk or custom format
Price (USD)
65.65
Online Exclusive
73.20
Save 7.55 (10%)
Each
Chemical Identifiers
CAS111-14-8
IUPAC Nameheptanoic acid
Molecular FormulaC7H14O2
InChI KeyMNWFXJYAOYHMED-UHFFFAOYSA-N
SMILESCCCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4205-1.4245 @ 20?C
CommentMaterial sourced in UK and US
Appearance (Color)Clear, colorless
FormLiquid
Identification (FTIR)Conforms (UK-sourced material)
View more
Heptanoic acid is used as an organic building block for the synthesis of a variety of chemical compounds. It is involved in the esterification of steroids and used in the preparation of active pharmaceutical ingredients such as trenbolone enanthate, testosterone enanthate, drostanolone enanthate and methenolone enanthate. It is also used as an internal standard during gas chromatography analysis of the butyl esters of volatile acids. Its ester derivatives are used in the fragrance and flavor industries as well as cosmetics and industrial lubricants in aviation, refrigeration and automobile. It is used to prepare sodium heptanoate, which is used as a corrosion inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Heptanoic acid is used as an organic building block for the synthesis of a variety of chemical compounds. It is involved in the esterification of steroids and used in the preparation of active pharmaceutical ingredients such as trenbolone enanthate, testosterone enanthate, drostanolone enanthate and methenolone enanthate. It is also used as an internal standard during gas chromatography analysis of the butyl esters of volatile acids. Its ester derivatives are used in the fragrance and flavor industries as well as cosmetics and industrial lubricants in aviation, refrigeration and automobile. It is used to prepare sodium heptanoate, which is used as a corrosion inhibitor.

Solubility
Miscible with acetone, ethanol, diethyl ether, dimethylformamide, dimethyl sulfoxide and nitric acid. Slightly soluble in carbon tetrachloride and water.

Notes
Incompatible with strong oxidizing agents, bases and reducing agents.
RUO – Research Use Only

General References:

  1. Lopez-Ruiz, J. A.; Pham, H. N.; Datye, A. K.; Davis, R. J. Reactivity and stability of supported Pd nanoparticles during the liquid-phase and gas-phase decarbonylation of heptanoic acid. Appl. Catal., A 2015, 504, 295-307.
  2. Aghzzaf, A. A.; Rhouta, B.; Rocca, E.; Khalil, A.; Caillet, C.; Hakkou, R. Heptanoic acid adsorption on grafted palygorskite and its application as controlled-release corrosion inhibitor of steel. Mater. Chem. Phys. 2014, 148 (1-2), 335-342.