2,3-Dibromopropionyl chloride, 95%
2,3-Dibromopropionyl chloride, 95%
2,3-Dibromopropionyl chloride, 95%
Thermo Scientific Chemicals

2,3-Dibromopropionyl chloride, 95%

CAS: 18791-02-1 | C3H3Br2ClO | 250.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A17712.30
also known as A17712-30
Price (USD)
397.65
Online Exclusive
442.00
Save 44.35 (10%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
397.65
Online Exclusive
442.00
Save 44.35 (10%)
Each
Chemical Identifiers
CAS18791-02-1
IUPAC Name2,3-dibromopropanoyl chloride
Molecular FormulaC3H3Br2ClO
InChI KeyHWKWYDXHMQQDQJ-UHFFFAOYNA-N
SMILESClC(=O)C(Br)CBr
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Methyl Ester GC)>94.0%
FormLiquid
Appearance (Color)Clear colorless to yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only