Amberlite IRA-410(Cl), ion exchange resin
Amberlite IRA-410(Cl), ion exchange resin
Amberlite IRA-410(Cl), ion exchange resin
Thermo Scientific Chemicals

Amberlite IRA-410(Cl), ion exchange resin

CAS: 9002-26-0
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number A17734.0E
also known as A17734-0E
Price (USD)
253.65
Special offer
Online exclusive
Ends: 28-Dec-2026
298.00
Save 44.35
Each
Quantity:
2500 g
Request bulk or custom format
Price (USD)
253.65
Special offer
Online exclusive
Ends: 28-Dec-2026
298.00
Save 44.35
Each
Chemical Identifiers
CAS9002-26-0
MDL NumberMFCD00132710
SpecificationsSpecification SheetSpecification Sheet
FormBeads
Harmonic mean size0.600-0.750mm
Moisture holding capacity45.0-51.0% (Cl-)
Particle Size>1180 micron: ≤ 5.0%
Appearance (Color)Pale yellow to pale orange
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only