2-Chlorobenzothiazole, 98%
2-Chlorobenzothiazole, 98%
2-Chlorobenzothiazole, 98%
Thermo Scientific Chemicals

2-Chlorobenzothiazole, 98%

CAS: 615-20-3 | C7H4ClNS | 169.626 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A17762.06
also known as A17762-06
Price (USD)
33.65
Online Exclusive
37.00
Save 3.35 (9%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
33.65
Online Exclusive
37.00
Save 3.35 (9%)
Each
Chemical Identifiers
CAS615-20-3
IUPAC Name2-chloro-1,3-benzothiazole
Molecular FormulaC7H4ClNS
InChI KeyBSQLQMLFTHJVKS-UHFFFAOYSA-N
SMILESClC1=NC2=CC=CC=C2S1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥98.0%
Refractive Index1.6360-1.6400 @ 20?C
Appearance (Color)Clear yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only