2-Chloro-m-xylene, 98%
2-Chloro-m-xylene, 98%
2-Chloro-m-xylene, 98%
Thermo Scientific Chemicals

2-Chloro-m-xylene, 98%

CAS: 6781-98-2 | C8H9Cl | 140.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A17867.22
also known as A17867-22
Price (USD)
206.65
Online Exclusive
229.00
Save 22.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
206.65
Online Exclusive
229.00
Save 22.35 (10%)
Each
Chemical Identifiers
CAS6781-98-2
IUPAC Name2-chloro-1,3-dimethylbenzene
Molecular FormulaC8H9Cl
InChI KeyVDXLAYAQGYCQEO-UHFFFAOYSA-N
SMILESCC1=CC=CC(C)=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.5260-1.5300 @ 20?C
It is employed as intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as intermediate for pharmaceutical.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in a tightly closed container in a cool, dry, well-ventilated area. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Jinkun Huang; and Steven P. Nolan. Efficient Cross-Coupling of Aryl Chlorides with Aryl Grignard Reagents (Kumada Reaction) Mediated by a Palladium/Imidazolium Chloride System. J. Am. Chem. Soc, 1999,121(42), 9889-9890.
  2. Karen E. Torraca; Xiaohua Huang; Cynthia A. Parrish; and Stephen L. Buchwald. An Efficient Intermolecular Palladium-Catalyzed Synthesis of Aryl Ethers. J. Am. Chem. Soc, 2001,123(43), 10770-10771.