Methyl glycolate, 98%
Methyl glycolate, 98%
Methyl glycolate, 98%
Thermo Scientific Chemicals

Methyl glycolate, 98%

CAS: 96-35-5 | C3H6O3 | 90.078 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A17870.22
also known as A17870-22
Price (USD)
312.65
Online Exclusive
347.00
Save 34.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
312.65
Online Exclusive
347.00
Save 34.35 (10%)
Each
Chemical Identifiers
CAS96-35-5
IUPAC Namemethyl 2-hydroxyacetate
Molecular FormulaC3H6O3
InChI KeyGSJFXBNYJCXDGI-UHFFFAOYSA-N
SMILESCOC(=O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
FormLiquid
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
Water Content (Karl Fischer Titration)≤1%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Methyl glycolate is a chemical reagent used in the synthesis of taxol and rapamycin. It is used in leather tanning and textile dyeing. It acts as a flavoring agent, preservative in food processing, in water treatment industries and as chemical intermediates. Further, it is used in emulsion polymers, ink and paint additive to improve flow properties and impart gloss. It plays an important role in producing triisopropylsilanyloxy-acetic acid methyl ester using imidazole as reagent.

Solubility
Miscible with water.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Wen, C.; Cui, Y.; Chen, X.; Zong, B.; Dai, W. L. Reaction temperature controlled selective hydrogenation of dimethyl oxalate to methyl glycolate and ethylene glycol over copper-hydroxyapatite catalysts. Appl. Catal., B 2015, 162, 483-493.
  2. Zhou, J.; Duan, X.; Ye, L.; Zheng, J.; Li, M. M. J.; Tsang, S. E.; Yuan, Y. Enhanced chemoselective hydrogenation of dimethyl oxalate to methyl glycolate over bimetallic Ag-Ni/SBA-15 catalysts. Appl. Catal., A 2015, 505, 344-353.