2,2,2-Trifluoroethyl p-toluenesulfonate, 98+%
2,2,2-Trifluoroethyl p-toluenesulfonate, 98+%
2,2,2-Trifluoroethyl p-toluenesulfonate, 98+%
Thermo Scientific Chemicals

2,2,2-Trifluoroethyl p-toluenesulfonate, 98+%

CAS: 433-06-7 | C9H9F3O3S | 254.223 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A17871.14
also known as A17871-14
Price (USD)
40.65
Special offer
Online exclusive
Ends: 30-Jun-2026
47.60
Save 6.95 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
40.65
Special offer
Online exclusive
Ends: 30-Jun-2026
47.60
Save 6.95 (15%)
Each
Chemical Identifiers
CAS433-06-7
IUPAC Name2,2,2-trifluoroethyl 4-methylbenzene-1-sulfonate
Molecular FormulaC9H9F3O3S
InChI KeyIGKCQDUYZULGBM-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)S(=O)(=O)OCC(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)37.0-43.0?C
Appearance (Color)White to pale cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only