DL-Malic acid, 98%
DL-Malic acid, 98%
DL-Malic acid, 98%
Thermo Scientific Chemicals

DL-Malic acid, 98%

CAS: 6915-15-7 | C4H6O5 | 134.087 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2500 g
Catalog number A17874.36
also known as A17874-36
Price (USD)
45.80
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
45.80
Each
Chemical Identifiers
CAS6915-15-7
IUPAC Name2-hydroxybutanedioic acid
Molecular FormulaC4H6O5
InChI KeyBJEPYKJPYRNKOW-UHFFFAOYNA-N
SMILESOC(CC(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Optical Rotation-0.5? to +0.5? (c= 8.5 in water)
Appearance (Color)White to cream
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (Silylated GC)≥97.5%
Identification (FTIR)Conforms
View more
DL-Malic acid is used as a food additive. It is a component of some artificial vinegar flavors. Further, it is used in the preparation of chiral compounds, including κ-opioid rece. It is utilized for chiral resolution by ligand-exchange capillary electrophoresis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Malic acid is used as a food additive. It is a component of some artificial vinegar flavors. Further, it is used in the preparation of chiral compounds, including κ-opioid rece. It is utilized for chiral resolution by ligand-exchange capillary electrophoresis.

Solubility
Soluble in acetone, ether, water, methanol and ethanol.

Notes
Incompatible with bases, oxidizing agents, reducing agents and alkali metals.
RUO – Research Use Only

General References:

  1. Zhang, Q.; Yang, Y.; Cheng, L.; Cao, C.; Ding, Z.; Wang, C.; Yang, W.; Hu, Y.; Li, Y. Thermodynamic models for determination of the solubility of dl-malic acid in methanol plus (acetonitrile, N,N-dimethylformamide, isopropyl alcohol) binary solvent mixtures. J. Chem. Thermodyn. 2015, 85, 148-154.
  2. Espinosa, K. C.; Cabrera, R. I. G.; López, A. B.; Roldán, M. A. T.; Cruz, N. A. V. Positive effect of reduced aeration rate on growth and stereospecificity of dl-malic acid consumption by Azospirillum brasilense: Improving the shelf life of a liquid inoculant formulation. J. Biotechnol. 2015, 195, 74-81.