Tin(II) oxide, 99%
Tin(II) oxide, 99%
Tin(II) oxide, 99%
Thermo Scientific Chemicals

Tin(II) oxide, 99%

CAS: 21651-19-4 | OSn | 134.71 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number A17889.0E
also known as A17889-0E
Price (USD)
560.65
Special offer
Online exclusive
Ends: 30-Jun-2026
659.00
Save 98.35 (15%)
Each
Quantity:
2500 g
Request bulk or custom format
Price (USD)
560.65
Special offer
Online exclusive
Ends: 30-Jun-2026
659.00
Save 98.35 (15%)
Each
Chemical Identifiers
CAS21651-19-4
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofATin (II): 87.0 to 87.5%
Elemental AnalysisLead, Pb ≤ 150ppm
Water Content (Karl Fischer Titration)≤0.1%
Appearance (Color)Dark grey to blue-black
Elemental AnalysisIron, Fe ≤ 60ppm
View more
Tin(II) oxide is used as reducing agent, soft abrasive, and in preparation of stannous salts. It is used in the manufacture of copper ruby glass, and for illumination with UV light.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tin(II) oxide is used as reducing agent, soft abrasive, and in preparation of stannous salts. It is used in the manufacture of copper ruby glass, and for illumination with UV light.

Solubility
Insoluble in water and alcohol.

Notes
Incompatible with materials like strong oxidizing agents, strong acids, strong reducing agents and alkali metals.
RUO – Research Use Only

General References:

  1. Yamaguchi, H.; Nakanishi, S.; Iba, H.; Itoh, T. Amorphous polymeric anode materials from poly(acrylic acid) and tin(II) oxide for lithium ion batteries. J. Power Sources 2015, 275, 1-5.
  2. Kaizra, S.; Louafi, Y.; Bellal, B.; Trari, M.; Rekhil, G. Electrochemical growth of tin(II) oxide films: Application in photocatalytic degradation of methylene blue. Mater. Sci. Semicond. Process. 2015, 30, 554-560.