4-Bromo-1,2-(methylenedioxy)benzene, 98%
4-Bromo-1,2-(methylenedioxy)benzene, 98%
4-Bromo-1,2-(methylenedioxy)benzene, 98%
4-Bromo-1,2-(methylenedioxy)benzene, 98%
Thermo Scientific Chemicals

4-Bromo-1,2-(methylenedioxy)benzene, 98%

CAS: 2635-13-4 | C7H5BrO2 | 201.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
Catalog number A17906.14
also known as A17906-14
Price (USD)
104.65
Online Exclusive
116.00
Save 11.35 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
104.65
Online Exclusive
116.00
Save 11.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS2635-13-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to pale brown
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.5810-1.5850 @ 20°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only