Tetramethylphosphonium bromide, 97%
Tetramethylphosphonium bromide, 97%
Tetramethylphosphonium bromide, 97%
Tetramethylphosphonium bromide, 97%
Thermo Scientific Chemicals

Tetramethylphosphonium bromide, 97%

CAS: 4519-28-2 | C4H12BrP | 171.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A17907.03
also known as A17907-03
Price (USD)
65.90
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
65.90
Each
Add to cart
Chemical Identifiers
CAS4519-28-2
IUPAC Nametetramethylphosphanium bromide
Molecular FormulaC4H12BrP
InChI KeyZTXFOCMYRCGSMU-UHFFFAOYSA-M
SMILES[Br-].C[P+](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow or pale pink-purple
FormCrystals or powder or crystalline powder or fused/lumpy solid
Assay (Titration ex Bromide)≥96.0 to ≤104.0%
It is used as a pharmaceutical intermediate and in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate and in chemical research.

Solubility
Insoluble in water.

Notes
Hygroscopic. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
RUO – Research Use Only

General References:

  1. Staffan Andersson and Susan Jagner. Coordination of Copper (∣) in Bis (tetramethylphosphonium) Tribromocuprate (∣) and Bis (tetraethylphosphonium) Di-u-bromo-dibromodicuprate (∣); Comparison od Anionic Configurations in Bromocuprates(∣) Crystallizing with Symmetrical Tetraalkylammonium and Related Cations. Acta chemica Scandinavica. Series A.,1987,41 230-236.
  2. Adel Rafai Far.; Alexander Shivanyuk.; and Julius Rebek, Jr. Water-Stabilized Cavitands.J. Am. Chem. Soc.,2002,124(12), 2854-2855.