Sodium thiosulfate pentahydrate, 99+%
Sodium thiosulfate pentahydrate, 99+%
Sodium thiosulfate pentahydrate, 99+%
Thermo Scientific Chemicals

Sodium thiosulfate pentahydrate, 99+%

CAS: 10102-17-7 | H10Na2O8S2 | 248.172 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10,000 g
500 g
2500 g
Catalog number A17914.0E
also known as A17914-0E
Price (USD)
99.30
Each
Quantity:
2500 g
Request bulk or custom format
Price (USD)
99.30
Each
Chemical Identifiers
CAS10102-17-7
IUPAC Namedisodium pentahydrate sulfanidesulfonate
Molecular FormulaH10Na2O8S2
InChI KeyPODWXQQNRWNDGD-UHFFFAOYSA-L
SMILESO.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or pellets
Assay (Redox Titration)≥99.0 to ≤101.0%
Identification (FTIR)Conforms
Appearance (Color)Colorless to white
Sodium thiosulfate pentahydrate is used as an analytical reagent in laboratories. It is used in photographic processing and in gold extraction. Its alkylated derivative S-alkylthiosulfonates are called Bunte salts. It is involved in the preparation of thioglycolic acid. Furthermore, it is used to dechlorinate tap water, for removing free bromine from solution and to check pH of bleach solutions with liquid indicators. Other used include tanning of leather, as an active component in hand warmers, in chemical heating pads and as an anionic surfactant to aid in dispersion of pharmaceutical preparations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium thiosulfate pentahydrate is used as an analytical reagent in laboratories. It is used in photographic processing and in gold extraction. Its alkylated derivative S-alkylthiosulfonates are called Bunte salts. It is involved in the preparation of thioglycolic acid. Furthermore, it is used to dechlorinate tap water, for removing free bromine from solution and to check pH of bleach solutions with liquid indicators. Other used include tanning of leather, as an active component in hand warmers, in chemical heating pads and as an anionic surfactant to aid in dispersion of pharmaceutical preparations.

Solubility
Soluble in water. Insoluble in alcohol.

Notes
Incompatible with sodium nitrate, acids, strong oxidizing agents, iodides, silver salts, mercury salts and lead.
RUO – Research Use Only

General References:

  1. Moral-Vico, J.; Sánchez-Redondo, S.; Lichtenstein, M. P.; Suñol, C.; Casañ-Pastor, N. Nanocomposites of iridium oxide and conducting polymers as electroactive phases in biological media. Acta Biomater. 2014, 10 (5), 2177-2186.
  2. James, T. H.; Cannon, C.; Apblett, A.; Materer, N. F. Sodium Dithionite Purity and Decomposition Products in Solid Samples Spanning 50 Years. Phosphorus Sulfur Silicon Relat. Elem. 2015, 190 (2), 158-169.