2,4-Difluorobenzoyl chloride, 97%
2,4-Difluorobenzoyl chloride, 97%
2,4-Difluorobenzoyl chloride, 97%
Thermo Scientific Chemicals

2,4-Difluorobenzoyl chloride, 97%

CAS: 72482-64-5 | C7H3ClF2O | 176.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A17920.06
also known as A17920-06
Price (USD)
33.65
Online Exclusive
37.40
Save 3.75 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
33.65
Online Exclusive
37.40
Save 3.75 (10%)
Each
Chemical Identifiers
CAS72482-64-5
IUPAC Name2,4-difluorobenzoyl chloride
Molecular FormulaC7H3ClF2O
InChI KeyJSWRVDNTKPAJLB-UHFFFAOYSA-N
SMILESFC1=CC=C(C(Cl)=O)C(F)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.5120-1.5170 @ 20?C
FormLiquid
Assay (GC)≥96.0%
2,4-Difluorobenzoyl chloride is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Difluorobenzoyl chloride is used as a pharmaceutical intermediate.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from water/ moisture, oxidizing agents, bases.
RUO – Research Use Only

General References:

  1. Christopher D. Gabbutt; John D. Hepworth; Michael W. J. Urquhart and Luis Millan Vazquez de Miguel. Synthesis and conjugate additions of2,3,4,9-tetrahydro-1H-xanthene-1,9-diones. J. Chem. Soc., Perkin Trans. 1. 1997, (12), 1819-1824.
  2. Andrzej Kapturkiewicz; Teng-Ming Chen; Inamur R Laskar; Jacek Nowacki. Electrochemiluminescence studies of the cyclometalated iridium(III) complexes with substituted 2-phenylbenzothiazole ligands. Electrochemistry Communications. 2004, 6, (8),827-831.