1-Heptylamine, 98+%
1-Heptylamine, 98+%
1-Heptylamine, 98+%
Thermo Scientific Chemicals

1-Heptylamine, 98+%

CAS: 111-68-2 | C7H17N | 115.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
50 g
250 g
Catalog number A17922.30
also known as A17922-30
Price (USD)
208.65
Special offer
Online exclusive
Ends: 30-Jun-2026
245.00
Save 36.35 (15%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
208.65
Special offer
Online exclusive
Ends: 30-Jun-2026
245.00
Save 36.35 (15%)
Each
Chemical Identifiers
CAS111-68-2
IUPAC Nameheptan-1-amine
Molecular FormulaC7H17N
InChI KeyWJYIASZWHGOTOU-UHFFFAOYSA-N
SMILESCCCCCCCN
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Identification (FTIR)Conforms
Refractive Index1.4230-1.4270 @ 20?C
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥98.0%
1-Heptylamine has been used as an internal standard in the determination of biogenic monoamines and biogenic diamines by the fluorescence-HPLC method. It has also been used to study the effect of hydration on the gaseous structure of protonated heptylamine by infrared photo dissociation (IRPD) spectroscopy and computational chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Heptylamine has been used as an internal standard in the determination of biogenic monoamines and biogenic diamines by the fluorescence-HPLC method. It has also been used to study the effect of hydration on the gaseous structure of protonated heptylamine by infrared photo dissociation (IRPD) spectroscopy and computational chemistry.

Solubility
Soluble in water 6791 mg/L @ 25°C.

Notes
Air sensitive. Store away from oxidizing agents, air, acids.
RUO – Research Use Only

General References:

  1. Yves Martin; David Boutin; Patrick Vermette. Study of the effect of process parameters for n-heptylamine plasma polymerization on final layer properties. Thin Solid Films. 1997, 515, (17), 6844-6852.
  2. Jian-Hua Sun; Rong-Ji Dai; Wei-Wei Meng; Yu-lin Deng. Efficient enzymatic kinetic resolution of 2-heptylamine with a highly active acyl donor. Catalysis Communications. 2010, 11, (11), 987-991.